Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Notch-1 Antibody (OTI3E12), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP273042IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273042IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273042IR Supplier Novus Biologicals Supplier No. NBP273042IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Notch-1 Monoclonal specifically detects Notch-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Notch-1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI3E12
Conjugate DyLight 755
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 2.1.2.11, EC 3.4.21.68, hN1, Notch (Drosophila) homolog 1 (translocation-associated), notch 1Notch homolog 1, translocation-associated (Drosophila), Notch homolog 1, translocation-associated, TAN1neurogenic locus notch homolog protein 1, Translocation-associated notch protein TAN-1
Gene Symbols NOTCH1
Host Species Mouse
Immunogen Recombinant protein expressed in E.coli corresponding to amino acids 2300-2556 of human Notch-1.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Apoptosis, Cancer Stem Cells, Golgi Apparatus Markers, Growth and Development, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 4851.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.