Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Notch-1 Rabbit anti-Mouse, Alexa Fluor 488, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB081998 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB081998 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB081998 Supplier Novus Biologicals Supplier No. NBP298331AF488

Rabbit Polyclonal Antibody

Notch-1 Polyclonal specifically detects Notch-1 in Mouse samples. It is validated for ELISA.

Specifications

Antigen Notch-1
Applications ELISA
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation 50 mM Sodium Borate
Gene Alias EC 2.1.2.11, EC 3.4.21.68, hN1, Notch (Drosophila) homolog 1 (translocation-associated), notch 1Notch homolog 1, translocation-associated (Drosophila), Notch homolog 1, translocation-associated, TAN1neurogenic locus notch homolog protein 1, Translocation-associated notch protein TAN-1
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Notch-1 (NP_032740.3; Met1-Gln526).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer Stem Cells, Golgi Apparatus Markers, Growth and Development, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 4851
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.