Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Notch-2 Antibody (605724), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB368373 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB368373 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB368373 Supplier Novus Biologicals Supplier No. FAB5196K
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Notch-2 Monoclonal specifically detects Notch-2 in Mouse samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen Notch-2
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 605724
Conjugate DyLight 488
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias AGS2, hN2, neurogenic locus notch homolog protein 2, Notch (Drosophila) homolog 2, notch 2Notch homolog 2 (Drosophila), Notch homolog 2
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse Notch-2, Leu26-Val528 (predicted), Accession # O35516
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Golgi Apparatus Markers, Growth and Development, Neuronal Cell Markers, Signal Transduction, Stem Cell Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 4853
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.