Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Notch-2 Antibody (B9) - Chimeric - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB001479 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001479 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001479 Supplier Novus Biologicals Supplier No. NBP2625600.025MG
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Notch-2 Monoclonal specifically detects Notch-2 in Human, Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence.

Specifications

Antigen Notch-2
Applications ELISA, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone B9
Conjugate Unconjugated
Dilution ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunofluorescence
Formulation PBS
Gene Alias AGS2, hN2, neurogenic locus notch homolog protein 2, Notch (Drosophila) homolog 2, notch 2Notch homolog 2 (Drosophila), Notch homolog 2
Gene Symbols NOTCH2
Host Species Rabbit
Immunogen NRR of mouse Notch 2.
Quantity 0.025 mg
Gene ID (Entrez) 4853
Test Specificity An engineered ScFv fragment antibody that binds to both mouse mNRR2 and human hNRR2 but not NRR1.
Target Species Human, Mouse
Content And Storage Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.