Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Notch-2 Antibody, Novus Biologicals™
SDP

Catalog No. NB60087901 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB60087901 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB60087901 Supplier Novus Biologicals Supplier No. NB6008790.1ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 4 publications

Notch-2 Polyclonal specifically detects Notch-2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Notch-2
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:400-1:2000, ELISA 1:30000-1:90000, Immunohistochemistry 1:200-1:800, Immunohistochemistry-Paraffin 1:500
Formulation 0.02M Potassium Phosphate, 0.15M Sodium Chloride, pH 7.2 with 0.1% Sodium Azide
Gene Alias AGS2, hN2, neurogenic locus notch homolog protein 2, Notch (Drosophila) homolog 2, notch 2Notch homolog 2 (Drosophila), Notch homolog 2
Gene Symbols NOTCH2
Host Species Rabbit
Immunogen Synthetic Peptide: C-NAAERTPSHSGHLQaa 2396-2409 (the total protein is 2471 aa).
Purification Method Unpurified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 4853
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.