Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Notch-3 Antibody (603532) [DyLight 650], Novus Biologicals™
SDP

Catalog No. NB399558 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB399558 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB399558 Supplier Novus Biologicals Supplier No. FAB1559W
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Notch-3 Monoclonal antibody specifically detects Notch-3 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen Notch-3
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 603532
Conjugate DyLight 650
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CADASIL, CASILneurogenic locus notch homolog protein 3, Notch (Drosophila) homolog 3, notch 3Notch homolog 3 (Drosophila), Notch homolog 3
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Notch-3, Ala40-Glu467, Accession # Q9UM47
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 4854
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.