Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NOTCH2 Monoclonal Antibody (16F11), PE, eBioscience™
GREENER_CHOICE

Catalog No. p-7176274
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-108-64 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-108-64 Supplier Invitrogen™ Supplier No. 12578682
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Description: This 16F11 monoclonal antibody reacts with the extracellular domain of human and mouse Notch2, one of four members of the Notch family of receptors. Notch receptors are 300-kDa single-pass transmembrane proteins. While the extracellular domain contains numerous epidermal growth factor-like repeats for ligand binding, the intracellular domain is involved in cell signaling. Upon binding its membrane-bound ligand (either Delta or Jagged), the Notch receptor undergoes proteolytic cleavage, first by ADAM-family metalloproteases and then by gamma-secretase. The second cleavage event releases the Notch intracellular domain (NICD), which subsequently translocates into the nucleus, heterodimerizes with the DNA-binding protein RBP-J, recruits co-activator molecules, and ultimately activates transcription. Notch2 is expressed on thymocytes (i.e., DN1 and DN2 subsets), activated peripheral T cells, and bone marrow. Moreover, expression of Notch2 has been reported to be altered in various cancers and lymphomas. In addition to its role in stem cell hematopoiesis, Notch2 plays a pivotal role in marginal zone B cell development. Notch2 also interacts with phosphorylated CREB1 to induce granzyme B transcription in cytotoxic T cells. Applications Reported: This 16F11 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 16F11 antibody has been tested by flow cytometric analysis of 24-hour anti-CD3-stimulated mouse splenocytes. This can be use...

Neurogenic locus notch homolog protein 2 (NOTCH2) is a highly conserved Notch signaling pathway protein. NOTCH2 is a type 1 transmembrane protein with an extracellular domain consisting of multiple epidermal growth factor-like (EGF) repeats, and an intracellular domain consisting of multiple, different domain types. NOTCH2 functions as a receptor for membrane-bound ligands Jagged1, Jagged2 and Delta1 to regulate cell-fate determination. Defects in NOTCH2 are the cause of Alagille syndrome type 2 (ALGS2). Alagille syndrome is an autosomal dominant multisystem disorder defined clinically by hepatic bile duct paucity and cholestasis in association with cardiac, skeletal, and ophthalmologic manifestations. There are characteristic facial features and less frequent clinical involvement of the renal and vascular systems.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOTCH2
Applications Flow Cytometry
Classification Monoclonal
Clone 16F11
Concentration 0.2 mg/mL
Conjugate PE
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene NOTCH2
Gene Accession No. O35516, Q04721
Gene Alias AGS2; AI853703; HJCYS; hN2; Motch B; N2; N2ECD; N2ICD; Neurogenic locus notch homolog protein 2; notch 2; Notch 2 extracellular truncation; Notch 2 intracellular domain; notch gene homolog 2; Notch homolog 2; notch receptor 2; Notch2
Gene Symbols NOTCH2
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18129, 4853
Target Species Human, Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.