Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ NotI (10 U/μL)

Catalog No. p-4521909
Click to view available options
Quantity:
1,500 units
300 units
500 units

The NotI restriction enzyme recognizes GC^GGCCGC sites and cuts best at 37°C in O buffer (Isoschizomers: CciNI).

Ad2 DNA digested with NotI, 0.7% agarose, 7 cleavage sites

conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

5'..G C▵G G C C G C...3'

3'..C G C C G G▵C G...5'

Conditions for 100% Activity:

  • 1X Buffer O: 50mM Tris-HCl (pH 7.5 at 37°C), 10mM MgCl2, 100mM NaCl and 0.1mg/mL BSA
  • Incubate at 37°C

Storage Buffer:

  • NotI is supplied in: 20mM Tris-HCl (pH 7.8 at 25°C), 100mM NaCl, 0.1mM EDTA, 1mM DTT, 0.02% Triton X-100, 0.2mg/mL BSA and 50% (v/v) glycerol

Ligation and Recleavage:

  • After 50-fold overdigestion with NotI, more than 95% of DNA fragments can be ligated and recut

Methylation Effects:

  • Dam: never overlaps —no effect
  • Dcm: never overlaps —no effect
  • CpG: completely overlaps —blocked
  • EcoKI: never overlaps —no effect
  • EcoBI: never overlaps —no effect

Digestion of Agarose-embedded DNA:

  • Minimum 5units of the enzyme are required for complete digestion of 1μg of agarose-embedded Adenovirus-2 DNA in 16 hours

Compatible Ends:

  • Bsp120I, CfrI, Eco52I

Note:

Assayed using pTZ19RJL2 plasmid DNA digested with BseLI. Supercoiled plasmids may require up to 5-fold more NotI for complete digestion than linear DNAs.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 10 U/μL
Methylation Sensitivity CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive
Enzyme Not I
Compatible Buffer 10X Buffer O
Sensitive to Heat Inactivation Yes
Optimal Reaction Temperature 37°C
Type IIS RE No
Quantity 1,500 units
Product Type Restriction Enzyme
Research Category Traditional Cloning
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.