Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOV/CCN3 Antibody (130), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB307857 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB307857 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB307857 Supplier Novus Biologicals Supplier No. NBP289369APC
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

NOV/CCN3 Monoclonal antibody specifically detects NOV/CCN3 in Human samples. It is validated for ELISA

Specifications

Antigen NOV/CCN3
Applications ELISA
Classification Monoclonal
Clone 130
Conjugate APC
Dilution ELISA
Formulation PBS
Gene Alias CCN family member 3, CCN3IGF-binding protein 9, IBP-9, IGFBP-9, IGFBP9novH, Insulin-like growth factor-binding protein 9, nephroblastoma overexpressed gene, Nephroblastoma-overexpressed gene protein homolog, NOVH, protein NOV homolog
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human NOV/CCN3 (Accession#: NP_002505.1; Met 1-Met 357).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4856
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.