Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ KOD Hot Start DNA Polymerase

Catalog No. 710865 Shop All MilliporeSigma Products
Click to view available options
Quantity:
1000 U
20 U
200 U

Catalog No. 71-086-5

Supplier: MilliporeSigma™ 710865

Only null left
Add to Cart
Add to Cart

For PCR amplification of long strand and GC-rich DNA templates, cloning and cDNA amplification applications

  • High accuracy, yield, and processivity of commercially available proofreading DNA polymerases
  • Amplifies genomic DNA templates up to 12 kbp
  • Amplifies plasmid and lambda DNA templates up to 21 kbp
  • Successfully amplifies GC-rich sequences
  • Eliminates mispriming and primer-dimer formation
  • Convenient room-temperature setup compatible with automation
  • Optimal KOD Hot Start Buffer for PCR performance over wide range of targets
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 U/μL
Content And Storage -20°C
Source Recombinant Thermococcus kodakaraensis KOD1 DNA polymerase expressed in E. coli
Quantity 20 U
Product Type KOD Hot Start DNA Polymerase

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.