Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ LB Agar, Miller

Catalog No. p-6921033 Shop All MilliporeSigma Products
Click to view available options
Quantity:
25 Easy

Used as granulated medium for cultivation of E. coli. MilliporeSigma™; Novagen™ LB Agar, Miller granules ensure rapid and uniform dissolution in water.

  • Prevents clumping of medium and inhalation of airborne powder
  • Supplied in several formats
  • EasyPak is aluminum foil pouch containing 37g granulated medium sufficient for 1L culture
  • Add EasyPak contents to 1L water, microwave to dissolve agar and sterilize
  • Supplied in 500g and 5kg bottles
  • Composition of LB Agar MILLER per liter is 5g yeast extract, 10g peptone from casein, 10g sodium chloride, and 12g agar-agar
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15°C to 30°C
Product Type Agar
Quantity 25 Easy
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.