Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ Origami™ B (DE3) Competent Cells

Catalog No. p-7020080 Shop All MilliporeSigma Products
Click to view available options
Quantity:
0.4 mL
1 mL

Origami B host strains carry the same mutations as the original Origami strain, except that they are derived from a lacZY mutant of BL21 to enable precise control of expression levels using IPTG.

Origami™ B host strains carry the same mutations in trxB and gor as the original Origami strains, except that they are derived from a lacZY mutant of BL21 to enable precise control of expression levels by adjusting the concentration of IPTG. Thus the Origami B strains combine the desirable characteristics of BL21, Tuner™, and Origami strains in one strain background. The mutations in trxB and gor are selectable on kanamycin and tetracycline, respectively; therefore, these strains cannot be used with plasmids that can only selected with kanamycin or tetracycline. These strains also include the lon and ompT deficiences of BL21, which increase protein stability. DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG.
TRUSTED_SUSTAINABILITY

Specifications

Efficiency Guaranteed efficiency: >2 x 106 cfu/μg
Content And Storage ≤-70°C
Quantity 1 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.