Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ Overnight Express™ Instant LB Medium

Catalog No. 717573 Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 Each
1 kg
5 Each

Used as granulated autoinduction culture medium for high-level protein production in pET and other IPTG-inducible bacterial expression systems. MilliporeSigma™; Novagen™ Overnight Express™ Instant LB Medium is a granulated medium ensures rapid, uniform dissolution in water, prevents clumping of the medium, and avoids inhalation of airborne powder.

  • Instant LB Medium is combination of Overnight Express Autoinduction System 1 components (Cat. No. 71300) with rich Luria Bertani (LB) broth and is supplied in two formats
  • EasyPak is aluminum foil pouch containing 45g granulated medium, sufficient for 1L culture
  • Add the EasyPak contents to 1L sterile water, supplement with 10mL glycerol, and microwave for 2 min to dissolve
  • Available in 1kg bottles
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15°C to 30°C
Product Type Microbiology Media
Packaging Type Glass Bottle
Quantity 1 Each
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.