Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EMD Millipore™ Novagen™ rLysozyme™ Solution Available on GSA/VA Contract for Federal Government customers only.

For lysis of Gram-negative bacteria

Manufacturer:  EMD Millipore™ 711104

 View more versions of this product

Catalog No. 71-110-4


  Check Availability
Add to cart

Disclaimers:

For Research Use Only


Specifications

Description

  • Contains highly purified and stabilized recombinant lysozyme
  • Catalyzes hydrolysis of N-acetylmuramide linkages in bacterial cell walls
  • Specific activity of rLysozyme (1700KU/mg) for E. coli lysis is 250 times greater than that of chicken egg white lysozyme
  • Optimally active at physiological pH
Description & Specifications

Specifications

Stabilized recombinant lysozyme
27 to 33KU/μL
1200KU
Supplied as a ready-to-use solution at a concentration range of 27 to 33KU/μL in 50% glycerol containing 50mM Tris-HCl, 0.1M NaCl, 0.1mM EDTA, 1 mM DTT, 0.1% TRITON™ X-100
7.5
-20°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit