Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ NOVAX™ Class 00 Electrical Safety Gloves Kit

Catalog No. 191682129 Shop All Protective Industrial Products Inc. Products
Click to view available options
Color:
Black
Orange
Size:
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Kit includes one pair of natural rubber electrical insulating gloves (150-00-11), a pair of top grain goatskin leather glove protectors (148-1000), and an 11 in. nylon glove bag.

  • Includes one pair of natural rubber electrical insulating gloves (150-00-11), a pair of top grain goatskin leather glove protectors (148-1000), and an 11 in. nylon glove bag
  • Class 00 insulating gloves will protect workers from up to 500 V
  • Meets NFPA 70E standards for use around arc flash hazards

Specifications

Certifications/Compliance ANSI/ASTM D120, Class 00, Type I, NFPA 70E
Applications Industrial
Color Black
Hand Specific
Material Natural Rubber, Top Grain Goatskin Leather
Product Type Electrical Insulating Gloves Kit
Size 9
For Use With (Application) Electrical Utility Work, Industrial Plant Maintenance, and Power Generation
Glove Cuff Style Straight
Includes Natural Rubber Electrical Insulating Gloves (150-00-11), Top Grain Goatskin Leather Glove Protectors (148-1000), and a Nylon Glove Bag (148-2136)
Series 150-SK-00
Length (English) 11 in.
Length (Metric) 28 cm
Voltage 500 V
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.