Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ iBind™ Cards

Catalog No. p-8394001
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 cards
40 cards
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SLF1010X4 40 cards
SLF1010 10 cards
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SLF1010X4 Supplier Invitrogen™ Supplier No. SLF1010X4
Only null left
Add to Cart
Add to Cart

Includes

Each box contains 10 cards

Requires

iBind Western Device

For use with the iBind Western Device

The automated iBind Western Device performs every step in the Western processing workflow from blocking to washes to antibody incubations. Each solution is released from the iBind device wells to the iBind Card through sequential lateral flow (SLF) where the solutions are wicked towards the stack region of the card. The glass fiber matrix of the card allows for homogenous and consistent flow of the solutions to the membrane, increasing the antigen-antibody interaction.

  • Unique, proprietary, glass-fiber matrix allows optimal and homogenous flow of immunodetection solutions
  • Long shelf life—store iBind Cards for 12 months at room temperature
  • Better antigen-antibody interaction—solutions flow between the iBind Card and membrane for maximum interaction between protein and antibody
  • Brings automation and simplicity to the Western protocol
  • Each iBind Card can run one mini-membrane

Proteins, Expression, Isolation and Analysis, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
Capacity 1 Mini Blot/Card
Detection Method Chromogenic, Chemiluminescence, Fluorescence
For Use With (Application) Western Blot
For Use With (Equipment) iBind Western Device
Membrane Compatibility Nitrocellulose, PVDF
Product Line iBind
Throughput 1 Mini Blot/Card
Type Card
Quantity 40 cards
Shipping Condition Room Temperature
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.