Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ iBlot™ Transfer Stack, PVDF, mini
GREENER_CHOICE

Catalog No. p-4919175
Encompass_Preferred
Click to view available options
Quantity:
10 sets
3 x 10 sets
Dimensions (LxW):
Mini, 8 cm x 8 cm

Includes

One box of iBlot Transfer Stacks containing 10 stacks

Used to transfer proteins with the iBlot Gel Transfer Device

The iBlot Transfer Stacks are used to transfer proteins using the iBlot Gel Transfer Device. The iBlot Transfer Stacks are disposable stacks that come in three varieties for different applications: iBlotGel Transfer Stacks have integrated PVDF or nitrocellulose transfer membranes to perform dry blotting of proteins; iBlot DNA Transfer Stacks have an integrated nylon transfer membrane to perform dry blotting of DNA, and iBlot Western Detection Stacks are used to perform Western detection on previously blotted nitrocellulose or PVDF membranes. Each iBlot Transfer Stack contains a copper electrode and appropriate cathode and anode buffers in the gel matrix to allow fast, reliable transfer of proteins or DNA.

PVDF membrane

  • 0.2μm pore size, low fluorescence
  • Has higher binding capacity than nitrocellulose
  • Preactivated and ready for use without any pretreatment with alcohols
  • Compatible with commonly used detection methods such as staining, immunodetection, fluorescence, or radiolabeling
  • Bind proteins due to hydrophobic interactions
  • Protein binding capacity: 240μg/cm2

Transfer stack sizes

  • The regular size (13 x 8.3cm) is used for all Novex midi-gel types (or equivalent) and E-PAGE™ 48 or 96 Gels
  • The mini size (8 x 8cm) is used for all Novex mini-gels (or equivalent)

Proteins, Expression, Isolation and Analysis, Western Blotting

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Length (Metric) 8 cm
Width (Metric) 8 cm
Quantity 3 x 10 sets
Format Transfer Stack
Material PVDF
Pore Size 0.2 μm
Content And Storage 30 sets of transfer stacks in 3 boxes. Store at room temperature.
Dimensions (LxW) Mini, 8 cm x 8 cm
Product Line iBlot
Sufficient For 30 Blots
Green Features Sustainable disposal
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.