Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Tropifluor™ PVDF Membrane, 0.45 μm pore size, 15 cm X 15 cm

Catalog No. T2234
Encompass_Preferred
Click to view available options
Quantity:
5 sheet(s)

Provide optimized results in western blotting applications with dioxetane-based substrate solutions (CSPD™ or CDP-Star™ substrates)

Provides the high binding and retention capacity that characterizes PVDF (polyvinylidene fluoride) membranes and high mechanical strength that permits convenient handling and withstands multiple re-probing. Low background and resulting superior signal-to-noise performance obtained with this membrane enables picogram detection levels of proteins in immunoblotting applications. Yields low background levels with use of Tropix chemiluminescent substrates and secondary antibody conjugates, enabling ultrasensitive detection. Recommended for immunoblotting applications where sensitivity is important.

  • Pore Size of 0.45 microns
  • Binding capacity: 125μg of protein/cm2
  • Size: 15 x 15cm

Proteins, Expression, Isolation and Analysis, Western Blotting

Order Info

Shipping Condition: Room Temperature

Warranty and Services

Guaranteed for 12 months from the date of purchase.

TRUSTED_SUSTAINABILITY

Specifications

Length (Metric) 15 cm
Width (Metric) 15 cm
Quantity 5 sheet(s)
Format Sheet
Material PVDF
Pore Size 0.45 μm
Content And Storage Membranes may be stored at room temperature.
Shipping Condition Room Temperature
Dimensions (LxW) 15 cm x 15 cm
Product Line Tropifluor

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.