Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ AbSelect™ Generic Antibody Purification Kit
Shop All R&D Systems Products

Click to view available options
Sufficient For:
1 Purification
3 Purifications
Description
The AbSelect method involves capture of the antibody on protein A resin and the removal of unwanted substances by a simple wash procedure. The purified product is then eluted and neutralized. The antibody to be purified or cleaned up ideally is in a volume of 100μL to 0.5mL, up to 500μg of antibody can be purified in each run.
Compatible with lightning-link antibody labeling system, which allows purified antibody to be rapidly conjugated with wide variety of labels
Recommended Storage
Store at 4°C. Do not freeze.
Specifications
Specifications
| Product Type | Antibody Purification Kit |
| Sufficient For | 3 Purifications |
| Content And Storage | 4°C. Do not freeze. |
| Quantity | 3 Tests |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction