Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ 5-Lipoxygenase Antibody Blocking Peptide
Shop All Novus Biologicals Products

Click to view available options
Quantity:
0.05 mg
Description
A peptide to ALOX5.
Specifications
Specifications
Gene ID (Entrez) | 240 |
Species | Human |
Content And Storage | Store at -80 C. Avoid freeze-thaw cycles. |
Formulation | Peptide dissolved in dH2O. Contains no BSA. |
For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB110-58749. For Further Blocking Peptide Related Protocol, Click Here . |
Gene Alias | 5-lipoxygenase, 5-LO, 5-LOX, 5 LPG, arachidonate 5-lipoxygenase, arachidonic 5-lipoxygenase alpha-10 isoform, arachidonic 5-lipoxygenase delta-10-13 isoform, arachidonic 5-lipoxygenase delta-13 isoform, arachidonic 5-lipoxygenase delta-p10 isoform, arachidonic acid 5-lipoxygenase, EC 1.13.11, EC 1.13.11.34, leukotriene A4 synthase, LOG5, MGC163204 |
Gene Symbol | ALOX5 |
Product Type | 5-Lipoxygenase |
Quantity | 0.05 mg |
Regulatory Status | RUO - research use only |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction