Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Annexin A8/ANXA8 Protein
SDP

Catalog No. p-5354401 Shop All R&D Systems Products
Click to view available options
Quantity:
0.1 mg
0.5 mg

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-327 of Human Annexin A8/ANXA8 The Recombinant Human Annexin A8/ANXA8 Protein is derived from E. coli. The Recombinant Human Annexin A8/ANXA8 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_001035173
For Use With (Application) ELISA, SDS-PAGE
Formulation Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 0.1M NaCl, 1mM DTT, 1mM EDTA, 10% glycerol
Gene ID (Entrez) 653145
Molecular Weight (g/mol) 39.0kDa
Purification Method Protein
Quantity 0.1 mg
Source Recombinant human ANXA8 protein, fused to His-tag at N-terminus, expressed in E. coli
Immunogen ANXA8, 1-327aa, Human, His tag, E. coli (NP_001035173).
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name Annexin A8/ANXA8
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.