Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ ARF6 Protein
Shop All Novus Biologicals Products

Click to view available options
Quantity:
0.1 mg
0.5 mg
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-175 of Human ARF6 The Recombinant Human ARF6 Protein is derived from E. coli. The Recombinant Human ARF6 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
Accession Number | NP_001654 |
Concentration | 1.0mg/mL |
For Use With (Application) | ELISA, SDS-PAGE |
Formulation | 20mM Tris-HCl buffer (pH 8.0) containing 2mM DTT, 20% glycerol, 0.2mM PMSF |
Gene ID (Entrez) | 382 |
Molecular Weight (g/mol) | 22.2kDa |
Purification Method | Protein |
Quantity | 0.1 mg |
Source | Recombinant human ARF6 protein, fused to His-tag at N-terminus, expressed in E. coli |
Immunogen | ARF6, 1-175aa, Human, His tag, E. coli (NP_001654). |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction