Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ CBFB Protein
Shop All Novus Biologicals Products

Click to view available options
Quantity:
0.05 mg
0.25 mg
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-182 of Human CBFB The Recombinant Human CBFB Protein is derived from E. coli. The Recombinant Human CBFB Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
Concentration | 0.5mg/mL |
For Use With (Application) | ELISA, SDS-PAGE |
Formulation | In 20mM MES (pH 6.0) containing 0.1mM PMSF, 10% Glycerol |
Gene ID (Entrez) | 865 |
Molecular Weight (g/mol) | 23.6kDa |
Purification Method | Protein |
Quantity | 0.05 mg |
Source | Human |
Immunogen | MGSSHHHHHH SSGLVPRGSH MPRVVPDQRS KFENEEFFRK LSRECEIKYT GFRDRPHEER QARFQNACRD GRSEIAFVAT GTNLSLQFFP ASWQGEQRQT PSREYVDLER EAGKVYLKAP MILNGVCVIW KGWIDLQRLD GMGCLEFDEE RAQQEDALAQ QAFEEARRRT REFEDRDRSH REEMEVRVSQ LLAVTGKKTT RP |
Storage Requirements | Store at -80°C. Avoid freeze-thaw cycles. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction