Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ CL-01 Cell Line
SDP

Catalog No. NBP149595 Shop All Novus Biologicals Products
Click to view available options
:
v6.5 Mouse embryonic stem cells

Useful to study mechanism of B cell differentiation and survival in vitro during antibody responses

  • Buffer: Approximately 3x106 cells in 1.5mL of freezing media and 10% DMSO
Applications: Studying gene regulation, function, and cellular and molecular events in B cells under normal or pathological conditions

Recommended Storage

Store in gas phase of liquid nitrogen

Warranty and Services

6 Months

Specifications

Content And Storage Store in gas phase of liquid nitrogen
For Use With (Application) Studying gene regulation, function, and cellular and molecular events in B cells under normal or pathological conditions
Product Type CL-01 Cell Line
Quantity 1.5mL

This product is for research use only and is not approved for use in humans or in clinical diagnosis

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.