Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Cystatin-9 Protein
Shop All R&D Systems Products

Click to view available options
Quantity:
100μg
Description
A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 29-159 of Human Cystatin-9 The Recombinant Human Cystatin-9 Protein is derived from E. coli. The Recombinant Human Cystatin-9 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Concentration | 1mg/mL |
| For Use With (Application) | SDS-PAGE |
| Formulation | Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 0.4M Urea, 10% glycerol |
| Molecular Weight (g/mol) | 17.2kDa |
| Purification Method | SDS-PAGE |
| Quantity | 100μg |
| Source | Human |
| Storage Requirements | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Regulatory Status | RUO |
| Common Name | Cystatin-9 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction