Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Fascin Protein
SDP

Catalog No. p-5354741 Shop All R&D Systems Products
Click to view available options
Quantity:
0.1 mg
0.5 mg

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-493 of Human Fascin The Recombinant Human Fascin Protein is derived from E. coli. The Recombinant Human Fascin Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_003079
Concentration 1.0mg/mL
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0) containing 20% glycerol, 2mM DTT, 100mM NaCl
Gene ID (Entrez) 6624
Molecular Weight (g/mol) 56.7kDa
Purification Method Protein
Quantity 0.1 mg
Source Recombinant human FSCN1 protein, fused to His-tag at N-terminus, expressed in E. coli
Immunogen FSCN1, 1-493aa, Human, His tag, E. coli (NP_003079).
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name Fascin
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.