Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ FCRLA/FCRLM1 Overexpression Lysate
SDP

Catalog No. NBL110666
Click to view available options
:
0.1mg
The FCRLA/FCRLM1 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot, SDS-Page.

Specifications

Cell Line The lysate was created in HEK293T cells, using plasmid ID RC21414 and based on accession number NM_32738. The protein contains a C-terminal DDK tag.
For Use With (Application) SDS-Page, Western Blot
Quantity 0.1 mg
Storage Buffer RIPA buffer
Purity Protein
Test Specificity Homo sapiens Fc receptor-like A (FCRLA), mRNA.
Content And Storage -80°C. Avoid freeze-thaw cycles.
Protein State Native

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.