Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ GMDS Protein
SDP

Catalog No. NBP17252110 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
250 μg
Formulation:
20mM Tris-HCl buffer (pH 8.0) containing 20% glycerol, 2mM DTT
20mM Tris-HCl buffer (pH 8.0) containing 30% glycerol, 0.1M NaCl, 1mM DTT, 0.1mM PMSF
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation
NBP17252110 100 μg 20mM Tris-HCl buffer (pH 8.0) containing 30% glycerol, 0.1M NaCl, 1mM DTT, 0.1mM PMSF
NBP17252225 250 μg 20mM Tris-HCl buffer (pH 8.0) containing 20% glycerol, 2mM DTT
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP17252110 Supplier Novus Biologicals™ Supplier No. NBP172521100UG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-372 of Human GMDS The Recombinant Human GMDS Protein is derived from E. coli. The Recombinant Human GMDS Protein has been validated for the following applications: SDS-Page.

Specifications

For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0) containing 30% glycerol, 0.1M NaCl, 1mM DTT, 0.1mM PMSF
Molecular Weight (g/mol) 44.1kDa
Purification Method SDS-PAGE
Quantity 100 μg
Source Human
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name GMDS
Conjugate Unconjugated
Form Liquid
Purity or Quality Grade >95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.