Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ HeLa Nuclear Cell Lysate
SDP

Catalog No. nb800pc9
Click to view available options
Quantity:
0.1 mg
The HeLa Nuclear Cell Lysate is derived from human. It has been validated for the following applications: Western Blot, SDS-Page.

Specifications

For Use With (Application) Western Blot, SDS-Page
Tissue Reproductive
Quantity 0.1 mg
Storage Buffer 1X SDS-PAGE Sample Buffer (62.5 mM Tris HCl, 2% SDS, 10% Glycerol and 0.005% bromophenol blue, pH 6.8), 10% (v/v) Glycerol
Purity Multi-step
Test Specificity HeLa cell nuclear extract lysate
Cellular Fraction Nuclear
Content And Storage Store at -70°C. Avoid freeze-thaw cycles.

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.