Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Histone H4 Peptide (non-modified a.a. 36-50), Biotin
Shop All R&D Systems Products

Click to view available options
Quantity:
0.5 mg
Description
non-modified a.a. 36-50 The Histone H4 Peptide has been validated for the following applications: ELISA, Functional, Gel Super Shift Assays, Radioimmunoassay, Peptide ELISA, Dot Blot, Block/Neutralize.
Specifications
Specifications
| Gene ID (Entrez) | 121504 |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| Formulation | Lyophilized. Reconstitute in diH20. |
| For Use With (Application) | Blocking, Neutralization, Dot Blot, ELISA, Gel Super Shift, Peptide ELISA, Radioimmunoassay |
| Product Type | Histone H4 |
| Quantity | 0.5 mg |
| Regulatory Status | RUO - research use only |
| Sequence | Biot-Ahx-RLARRGGVKRISGLI-amide |
| Termination | Biotin |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction