Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Histone H4 Peptide (non-modified a.a. 36-50), Biotin
SDP

Catalog No. p-5354212 Shop All R&D Systems Products
Click to view available options
Quantity:
0.5 mg

Highly purified. Generating reliable and reproducible results.

non-modified a.a. 36-50 The Histone H4 Peptide has been validated for the following applications: ELISA, Functional, Gel Super Shift Assays, Radioimmunoassay, Peptide ELISA, Dot Blot, Block/Neutralize.

Specifications

Gene ID (Entrez) 121504
Content And Storage Store at -80°C. Avoid freeze-thaw cycles.
Formulation Lyophilized. Reconstitute in diH20.
For Use With (Application) Blocking, Neutralization, Dot Blot, ELISA, Gel Super Shift, Peptide ELISA, Radioimmunoassay
Product Type Histone H4
Quantity 0.5 mg
Regulatory Status RUO - research use only
Sequence Biot-Ahx-RLARRGGVKRISGLI-amide
Termination Biotin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.