Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ HMGB1/HMG-1 Protein
SDP

Catalog No. p-5354844 Shop All R&D Systems Products
Click to view available options
Quantity:
0.1 mg
0.5 mg

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: ELISA, Immunoassay, Bioactivity

Recombinant protein corresponding to Met1-Glu215 of mouse HMGB-1. Accession #: NM_010439. Source: E.coli The Recombinant Mouse HMGB1/HMG-1 Protein is derived from E. coli. The Recombinant Mouse HMGB1/HMG-1 Protein has been validated for the following applications: ELISA, Immunoassay, Bioactivity.

Specifications

For Use With (Application) ELISA, Immunoassay
Formulation Sterile liquid; 20mM Tris-HCl, pH 8.0, 0.3M NaCl, 1mM DTT. 0.22μm filtered.
Quantity 0.1 mg
Source Mouse
Immunogen HMGB1/HMG-1
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name The recombinant mouse HMGB-1 has been tested by bioassay, yielding ED50 of 3μg/mL.
Conjugate Unconjugated
Recombinant Recombinant
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.