Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Human DECR2 His Protein
Shop All R&D Systems Products

Click to view available options
:
50μg; Unlabeled
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-292 of Human DECR2 The Recombinant Human DECR2 Protein is derived from E. coli. The Recombinant Human DECR2 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Accession Number | Q9NUI1 |
| For Use With (Application) | SDS-PAGE |
| Formulation | Liquid. 20mM Tris-HCl buffer (pH8.0) containing 40% glycerol, 0.15M NaCl, 1mM DTT |
| Gene ID (Entrez) | 26063 |
| Molecular Weight (g/mol) | 33.2kDa |
| Name | DECR2 Protein |
| Purification Method | SDS-PAGE |
| Storage Requirements | 4°C short term, −20°C long term. Avoid freeze-thaw cycles. |
| Cross Reactivity | Human |
| Purity or Quality Grade | >95% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction