Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Human Glutaredoxin 2 His Protein
Shop All Novus Biologicals Products

Click to view available options
:
0.1mg; Unlabeled
Description
A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 20-164 of Human Glutaredoxin 2 The Recombinant Human Glutaredoxin 2 Protein is derived from E. coli. The Recombinant Human Glutaredoxin 2 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
Accession Number | NP_932066 |
For Use With (Application) | ELISA, Western Blot |
Formulation | 20mM Tris-HCl buffer (pH 8.0), 0.1mM PMSF, 10% glycerol |
Gene ID (Entrez) | 51022 |
Molecular Weight (g/mol) | 17kDa |
Name | Glutaredoxin 2 Protein |
Purification Method | Protein |
Immunogen | MSAGWLDRAA GAAGAAAAAA SGMESNTSSS LENLATAPVN QIQETISDNC VVIFSKTSCS YCTMAKKLFH DMNVNYKVVE LDLLEYGNQF QDALYKMTGE RTVPRIFVNG TFIGGATDTH RLHKEGKLLP LVHQCYLKKS KRKEFQLEHH HHHH |
Storage Requirements | −80°C. Avoid freeze-thaw cycles. |
Cross Reactivity | Human |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction