Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Human HADH His Protein
Shop All Novus Biologicals Products

Click to view available options
:
0.1mg; Unlabeled
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 13-314 of Human HADH The Recombinant Human HADH Protein is derived from E. coli. The Recombinant Human HADH Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
Accession Number | AAH00306 |
For Use With (Application) | ELISA, SDS-PAGE |
Formulation | 20mM Tris-HCl buffer (pH8.0), 20% glycerol, 0.1 M NaCl |
Gene ID (Entrez) | 3033 |
Molecular Weight (g/mol) | 35.1kDa |
Name | HADH Protein |
Purification Method | Protein |
Immunogen | MGSSHHHHHH SSGLVPRGSH MSSSSTASAS AKKIIVKHVT VIGGGLMGAG IAQVAAATGH TVVLVDQTED ILAKSKKGIE ESLRKVAKKK FAENPKAGDE FVEKTLSTIA TSTDAASVVH STDLVVEAIV ENLKVKNELF KRLDKFAAEH TIFASNTSSL QITSIANATT RQDRFAGLHF FNPVPVMKLV EVIKTPMTSQ KTFESLVDFS KALGKHPVSC KDTPGFIVNR LLVPYLMEAI RLYERGDASK EDIDTAMKLG AGYPMGPFEL LDYVGLDTTK FIVDGWHEMD AENPLHQPSP SLNKLVAENK FGKKTGEGFY KYK |
Storage Requirements | −80°C. Avoid freeze-thaw cycles. |
Cross Reactivity | Human |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction