Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Muscle Phosphofructokinase/PFKM/PFK-1 His Protein
SDP

Catalog No. nbp144494 Shop All R&D Systems Products
Click to view available options
:
0.1mg; Unlabeled

Catalog No. NBP144494

Supplier: Novus Biologicals™ NBP144494

Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-780 of Human Muscle Phosphofructokinase/PFKM/PFK-1 The Recombinant Human Muscle Phosphofructokinase/PFKM/PFK-1 Protein is derived from E. coli. The Recombinant Human Muscle Phosphofructokinase/PFKM/PFK-1 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_000280
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH8.0), 5mM DTT, 0.2 M NaCl, and 20% glycerol
Gene ID (Entrez) 5213
Molecular Weight (g/mol) 87.3kDa
Name Muscle Phosphofructokinase/PFKM/PFK-1 Protein
Purification Method Protein
Immunogen MGSSHHHHHH SSGLVPRGSH MTHEEHHAAK TLGIGKAIAV LTSGGDAQGM NAAVRAVVRV GIFTGARVFF VHEGYQGLVD GGDHIKEATW ESVSMMLQLG GTVIGSARCK DFREREGRLR AAYNLVKRGI TNLCVIGGDG SLTGADTFRS EWSDLLSDLQ KAGKITDEEA TKSSYLNIVG LVGSIDNDFC GTDMTIGTDS ALHRIMEIVD AITTTAQSHQ RTFVLEVMGR HCGYLALVTS LSCGADWVFI PECPPDDDWE EHLCRRLSET RTRGSRLNII IVAEGAIDKN GKPITSEDIK NLVVKRLGYD TRVTVLGHVQ RGGTPSAFDR ILGSRMGVEA VMALLEGTPD TPACVVSLSG NQAVRLPLME CVQVTKDVTK AMDEKKFDEA LKLRGRSFMN NWEVYKLLAH VRPPVSKSGS HTVAVMNVGA PAAGMNAAVR STVRIGLIQG NRVLVVHDGF EGLAKGQIEE AGWSYVGGWT GQGGSKLGTK RTLPKKSFEQ ISANITKFNI QGLVIIGGFE AYTGGLELME GRKQFDELCI PFVVIPATVS NNVPGSDFSV GADTALNTIC TTCDRIKQSA AGTKRRVFII ETMGGYCGYL ATMAGLAAGA DAAYIFEEPF TIRDLQANVE HLVQKMKTTV KRGLVLRNEK CNENYTTDFI FNLYSEEGKG IFDSRKNVLG HMQQGGSPTP FDRNFATKMG AKAMNWMSGK IKESYRNGRI FANTPDSGCV LGMRKRALVF QPVAELKDQT DFEHRIPKEQ WWLKLRPILK ILAKYEIDLD TSDHAHLEHI TRKRSGEAAV
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Cross Reactivity Human
Purity or Quality Grade >90%, by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.