Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human PPP1R11 His Protein
SDP

Catalog No. nbp223379 Shop All R&D Systems Products
Click to view available options
Quantity:
0.05 mg

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-126 of Human PPP1R11 The Recombinant Human PPP1R11 Protein is derived from E. coli. The Recombinant Human PPP1R11 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_068778
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.2 M NaCl, 20% glycerol, 1mM DTT
Gene ID (Entrez) 6992
Molecular Weight (g/mol) 16.3kDa
Name PPP1R11 Protein
Purification Method SDS-PAGE
Quantity 0.05 mg
Source E. Coli
Immunogen PPP1R11, 1-126aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMAEAGAG LSETVTETTV TVTTEPENRS LTIKLRKRKP EKKVEWTSDT VDNEHMGRRS SKCCCIYEKP RAFGESSTES DEEEEEGCGH THCVRGHRKG RRRATLGPTP TTPPQPPDPS QPPPGPMQH
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category Signal Transduction
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.