Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Human RBP4/Retinol-Binding Protein Protein
Shop All R&D Systems Products

Click to view available options
:
0.02mg; Unlabeled
Description
Recombinant bioactive protein for Human RBP4/Retinol-Binding Protein 4 The Recombinant Human RBP4/Retinol-Binding Protein 4 Protein is derived from E. coli. The Recombinant Human RBP4/Retinol-Binding Protein 4 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Accession Number | P02753 |
| Formulation | Sterile filtered solution (1mg/mL ) in PBS, pH 7.4 |
| Gene ID (Entrez) | 5950 |
| Name | RBP4/Retinol-Binding protein Protein |
| Purification Method | SDS-PAGE |
| Immunogen | Amino acid Sequence: MERDCRVSSF RVKENFDKAR FSGTWYAMAK KDPEGLFLQD NIVAEFSVDE TGQMSATAKGRVRLLNNWDV CADMVGTFTD TEDPAKFKMK YWGVASFLQK GNDDHWIVDT DYDTYAVQYS CRLLNLDGTC ADSYSFVFSR DPNGLPPEAQ KIVRQRQEEL CLARQYRLIV HNGYCDGRSE RNLL |
| Storage Requirements | −20°C. Avoid freeze-thaw cycles. |
| Cross Reactivity | Human |
| Purity or Quality Grade | >95% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction