Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human STYX His Protein
SDP

Catalog No. NBP19907650 Shop All R&D Systems Products
Click to view available options
:
50μg; Unlabeled

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-223 of Human STYX The Recombinant Human STYX Protein is derived from E. coli. The Recombinant Human STYX Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number Q8WUJ0
For Use With (Application) SDS-PAGE
Formulation Liquid. 20mM Tris-HCl buffer (pH8.0) containing 30% glycerol 1mM DTT,0.1M NaCl
Gene ID (Entrez) 6815
Molecular Weight (g/mol) 28kDa
Name STYX Protein
Purification Method SDS-PAGE
Storage Requirements 4°C short term, −20°C long term. Avoid freeze-thaw cycles.
Cross Reactivity Human
Purity or Quality Grade >85%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.