Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Human TANK Protein
Shop All Novus Biologicals Products

Click to view available options
:
0.1mg; Unlabeled
Description
An un-tagged recombinant protein corresponding to the amino acids 1-119 of Human TANK The Recombinant Human TANK Protein is derived from E. coli. The Recombinant Human TANK Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
Accession Number | NP_597841 |
For Use With (Application) | ELISA, SDS-PAGE |
Formulation | 20mM Tris-HCl buffer (pH8.0) |
Gene ID (Entrez) | 10010 |
Molecular Weight (g/mol) | 13.6kDa |
Name | TANK Protein |
Purification Method | Protein |
Immunogen | MDKNIGEQLN KAYEAFRQAC MDRDSAVKEL QQKTENYEQR IREQQEQLSL QQTIIDKLKS QLLLVNSTQD NNYGCVPLLE DSETRKNNLT LDQPQDKVIS GIAREKLPKV DIASAESSI |
Storage Requirements | −80°C. Avoid freeze-thaw cycles. |
Cross Reactivity | Human |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction