Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human VEGF Protein
SDP

Catalog No. nbc121277 Shop All R&D Systems Products
Click to view available options
:
0.1mg; Unlabeled

Highly purified and high bioactivity. Generating reliable and reproducible results.

Human VEGF121 stimulates cell proliferation of the HUVEC human umbilical vein endothelial cells. The ED50 range less than or equal to 4 ng/ml.

Specifications

Accession Number NP_001020541
For Use With (Application) Bioactivity, In vitro Assay, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 10 % glycerol
Gene ID (Entrez) 7422
Molecular Weight (g/mol) 14.2kDa
Name VEGF Protein
Purification Method Affinity purified
Immunogen MAPMAEGGGQ NHHEVVKFMD VYQRSYCHPI ETLVDIFQEY PDEIEYIFKP SCVPLMRCGG CCNDEGLECV PTEESNITMQ IMRIKPHQGQ HIGEMSFLQH NKCECRPKKD RARQEKCDKP RR
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Cross Reactivity Human
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.