Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Lightning-Link (R) Alkaline Phosphatase Antibody Labeling Kit
SDP

Catalog No. p-7070035 Shop All R&D Systems Products
Click to view available options
Quantity:
1 mg
100 μg
30 μg
300 μg

Catalog No. 70-200-05

Supplier: Novus Biologicals™ 7020005

Only null left
Add to Cart
Add to Cart

Applications: ELISA

Alkaline phosphatase (AP) is a hydrolyase enzyme that is frequently conjugated to antibodies for use in immunoassays. PNPP is a popular colorimetric substrate for AP; the product of the reaction, p-nitrophenol, can be read at 405nM. Fluorogenic substrates (e.g. 4-methylumbelliferyl phosphate; MUP) may also be employed.

Order Info

Quantity

Specifications

Conjugate Alkaline Phosphatase
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 100 μg
Kit Contents Glass vial(s) of Lightning-LinkTM mix (1 or 3 vials, depending on pack size), 1 vial of LL-AP Modifier reagent, 1 vial of LL-AP Quencher reagent
Product Type Alkaline Phosphatase Antibody Labeling Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.