Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Lightning-Link (R) APC-Cy5.5 Antibody Labeling Kit
SDP

Catalog No. p-7070050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
30 μg
300 μg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-400-10 300 μg
76-400-05 100 μg
76-400-15 1 mg
76-400-30 30 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 76-400-10 Supplier Novus Biologicals™ Supplier No. 7640010
Only null left
Add to Cart
Add to Cart

Antibody labeling kit

The antibody is covalently bonded to the label in a directional and controlled process at near-neutral pH. APC/Cy5.5 is a tandem conjugate. In the Lightning-Link APC/Cy5.5 the tandem is excited at 633nm, wavelength at which the APC has its maximum absorption. The excited APC functions then as an energy donor, where the energy is transferred via FRET (Fluorescence Resonance Energy Transfer) to the Cy5.5. The Cy5.5 emits then the energy received from the APC in the form of long wavelength light at 694nm. The amount of antibody used for labeling should be the same as the pack size of LL-APC-Cy5.5. For any new antibody, optimization of the ratio of antibody to dye is often worthwhile. If your antibody needs to be concentrated, purified, or cleaned up, consider purchasing an antibody clean up kit.

Order Info

Quantity

Specifications

Quantity 300 μg
Conjugate APC/Cy5.5
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Kit Contents Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL Modifier reagent, 1 vial of LL Quencher FD reagent
Product Type APC-Cy5.5 Antibody Labeling Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.