Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Lightning-Link (R) B-Phycoerythrin Antibody Labeling Kit
SDP

Catalog No. 7160015 Shop All Novus Biologicals Products
Click to view available options
Quantity:
180 μg
60 μg
600 μg

Applications: Flow Cytometry

B-Phycoerythrin (B-PE) is a fluorescent protein from phycobiliprotein family. It has a primary absorption peak at 546 nm and a secondary peak at 563 nm. When excited at 546 nm it has a strong fluorescence emission with its maximum at 575 nm.

Order Info

Quantity

Specifications

Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 600 μg
Conjugate B-Phycoerythrin
Kit Contents Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL-B-PE Modifier reagent, 1 vial of LL-B-PE Quencher reagent
Product Type B-Phycoerythrin Antibody Labeling Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.