Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Lightning-Link (R) HRP Antibody Labeling Kit
Shop All R&D Systems Products

Click to view available options
Quantity:
1 mg
3 x 10 μg Reaction
3 x 100 μg Reaction
5 mg
5 x 1 mg Reaction
Description
Horseradish peroxidase is a 44kDa glycoprotein with 6 lysine residues which can be conjugated to labeled biomolecules using Lightning-Link. The enzyme label can be visualized by chromogenic reactions; for example diaminobenzidine (DAB) in the presence of hydrogen peroxide (H202) is converted in to a water insoluble brown pigment. Other substrates which can be used to measure horseradish peroxidase activity include ABTS, TMB and TMBUS.
Order Info
Quantity
Specifications
Specifications
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Quantity | 3 x 10 μg Reaction |
| Conjugate | HRP |
| Kit Contents | Glass vial(s) of Lightning-Link mix (1, 3 or 5 vials, depending on pack size), 1 vial of LL Modifier reagent, 1 vial of LL Quencher reagent |
| Product Type | HRP Antibody Labeling Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction