Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Lightning-Link (R) PerCP-Cy5.5 Antibody Labeling Kit
SDP

Catalog No. p-7070049 Shop All Novus Biologicals Products
Click to view available options
Quantity:
1 mg
100 μg
30 μg
300 μg

Applications: Flow Cytometry, Electron Microscopy, Flow (Cell Surface)

PerCP/Cy5.5 is a tandem conjugate. The PerCP is excited at 482nm and functions as an energy donor for the Cy5.5. Energy is transferred from the PerCP to the Cy5.5 via energy resonance transfer. The Cy5.5 emits the energy received from the PerCP in the form of long wavelength light at 700nm.

Order Info

Quantity

Specifications

Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 100 μg
Conjugate PerCP/Cy5.5
Kit Contents Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL Modifier reagent, 1 vial of LL Quencher FD reagent
Product Type PerCP-Cy5.5 Antibody Labeling Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.