Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Lightning-Link (R) R-PE Antibody Labeling Kit
SDP

Catalog No. p-7070036 Shop All R&D Systems Products
Click to view available options
Quantity:
1 mg Reaction
1 x 100 μg Reaction
1 x 5 mg Reaction
3 x 60 μg Reaction
5 x 1 mg Reaction

Catalog No. 70-300-03

Supplier: Novus Biologicals™ 7030003

Only null left
Add to Cart
Add to Cart

Applications: Flow Cytometry

R-Phycoerythrin (R-PE) is a fluorescent protein from the phycobiliprotein family, and is isolated from red algae. The absorbance spectrum of R-PE covers a broad range of excitation wavelengths, which provides an advantage for multi-color immunofluorescent staining or cell sorting. R-PE is one of the most intensely fluorescent phycobiliprotein having orange fluorescence. It is significantly brighter and more photostable than conventional organic fluorophores and it has a high quantum yield.

Order Info

Quantity

Specifications

Quantity 5 x 1 mg Reaction
Content And Storage Store at -20°C in the dark. Avoid freeze-thaw cycles.
Conjugate PE
Product Type R-PE Antibody Labeling Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.