Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Lightning-Link (R) Rapid Atto390 Antibody Labeling Kit
Shop All R&D Systems Products

Click to view available options
Quantity:
30 μg
300 μg
Description
Atto390 is one of a new generation of fluorescent labels with a coumarin structure. It has a strong absorption at 390nm, high fluorescence at 479nm (extinction coefficient 2.4 x10 cm&superscript_begin;-1&superscript_end;M&superscript_begin;-1&superscript_end;) and high quantum yield.
Order Info
Quantity
Specifications
Specifications
| Conjugate | Atto 390 |
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Quantity | 300 μg |
| Kit Contents | Glass vial(s) of Lightning-Link™ Rapidmix (1 or 3 vials, depending on pack size), 1 vial of LL-Rapid Modifier reagent, 1 vial of LL-Rapid Quencher reagent |
| Product Type | Rapid Atto390 Antibody Labeling Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction