Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ LOX propeptide Antibody Blocking Peptide
Shop All R&D Systems Products

Click to view available options
Quantity:
0.05 mg
Description
A peptide to LOX.
Specifications
Specifications
| Gene ID (Entrez) | 4015 |
| Species | Human |
| Accession Number | 4015 |
| Content And Storage | Store at -80 C. Avoid freeze-thaw cycles. |
| Formulation | Peptide dissolved in dH2O. Contains no BSA. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NBP1-30327. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | lysyl oxidaseEC 1.4.3.13, MGC105112, protein-lysine 6-oxidase |
| Gene Symbol | LOX |
| Product Type | LOX propeptide |
| Quantity | 0.05 mg |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction