Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ LSM1 Protein
Shop All R&D Systems Products

Click to view available options
Quantity:
0.1 mg
0.5 mg
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-133 of Human LSM1 The Recombinant Human LSM1 Protein is derived from E. coli. The Recombinant Human LSM1 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Concentration | 1mg/mL |
| For Use With (Application) | ELISA, SDS-PAGE |
| Formulation | Liquid. In 20mM Tris-HCl buffer (pH 7.5) containing 1mM DTT, 10% glycerol, 0.1M NaCl |
| Gene ID (Entrez) | 27257 |
| Molecular Weight (g/mol) | 17.3kDa |
| Purification Method | Protein |
| Quantity | 0.1 mg |
| Source | Human |
| Immunogen | LSM1, 1- 133 aa. MGSSHHHHHHSSGLVPRGSHMNYMPGTASLIEDIDKKHLVLLRDGRTLIGFLRSIDQFANLVLHQTVERIHVGKKYGDIPRGIFVVRGENVVLLGEIDLEKESDTPLQQVSIEEILEEQRVEQQTKLEAEKLKVQALKDRGLSIPRADTLDEY |
| Storage Requirements | Store at -80°C. Avoid freeze-thaw cycles. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction